Learn More
United Scientific™ Acrylic Demonstration Lens Set
SDPSCI_ED
Catalog No. S42920 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Diameter (Metric) Lens:
38 mm
50 mm
75 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Lens
S42920 38 mm
S42923 75 mm
S139466 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S42920 Supplier United Scientific™ Supplier No. LSTA38
Add to Cart
Add to Cart
Set of six lenses of a variety of types.
  • Each single-diameter set includes a double convex, plano-convex, concavo-convex, double concave, plano-concave and convexo-concave lens
  • Three diameters available
  • Includes an activity guide
Specifications
Product Type Lens Set
For Use With (Application) Teaching and demonstrating the basic principles of optics
Includes 6 Lenses, one each of double convex, plano convex, concavo convex, double concave, plano concave, and convexo concave lens, activity guide and a reusable cardboard storage box
Quantity 6 Pk.
Diameter (English) Lens 1.5 in.
Diameter (Metric) Lens 38 mm