Learn More
Electron Microscopy Sciences Acrylic Desiccator Locking Cabinet with Hygrometer, 16" x 9", 10-1/2" height
SDP
Supplier:  Electron Microscopy Sciences 7122240
Encompass_Preferred
For dry, dust-free storage at temperatures up to 120 degree F. They can be used as incubators, freezers, and refrigerators or at room temperature. Cabinets have airtight doors that need no lubrication. Sliding perforated shelves for easy access; transparent acrylic walls for see-throguh storage. The third bottom shelf accommodates desiccants. Cabinets are also available with locks and a hygrometer.
Hygrometer has a temperature range of -58 degree F to 158 degree F with +/-1.5 degree F accuracy up to 104 degree F. Humidity range is 20 to 95% with +/-5% accuracy. LCD display. Includes 2 'AAA' batteries. Hygrometer temperature range is -58 to 158 deg F.
Footprint: 16" x 9"
Height: 10-1/2"
Catalog No. 50-930-921