Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Acrylic Equilateral Prism
SDPSCI_ED

Catalog No. S15760 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (Metric):
50 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S15760 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15760 Supplier United Scientific™ Supplier No. PAE050
Only null left
Add to Cart
Add to Cart

Ideal for classroom use.

  • This equilateral prism is ideal for lessons on optics, wavelengths and more.
  • Made from quality clear acrylic sheet with highly polished surfaces
  • Available in six sizes

Specifications

Material Acrylic
Product Type Prism
For Use With (Application) Optics and Light
Length (Metric) 50 mm
Length (English) 1.96 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.