Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Acrylic Storage Modules for Sample Cups
SCI_ED

Catalog No. S29193 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
100 Sample Cups
Storage Holder: 14 810 67
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S29193 Storage Holder: 14 810 67
S29192 100 Sample Cups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S29193

missing translation for 'validMainframeMsg'

Storage for hundreds of 13mm sample cups in less than a square foot of space.

  • Clear acrylic shelves with white base
  • Solid cover available separately to protect top holder
  • Up to six holders can be stacked under one solid cover

Order Info

Racks with colored bases are available on special order at slightly higher prices. Contact your Fisher Scientific Customer Service Representative for ordering information.

Specifications

Type Storage Module
Material Acrylic
Color Clear
Description Storage rack for sample cups
For Use With (Equipment) Storage Holder: 14 810 67
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.