Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acryloyl Chloride, Stabilized 98%, Thermo Scientific™

Catalog No. AC102780010
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC102780010 1 L
AC102782500 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102780010 Supplier Thermo Scientific Chemicals Supplier No. 102780010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-68-6
Molecular Formula C3H3ClO
Molecular Weight (g/mol) 90.51
MDL Number MFCD00000731
InChI Key HFBMWMNUJJDEQZ-UHFFFAOYSA-N
Synonym acryloyl chloride, acrylyl chloride, 2-propenoyl chloride, acrylic acid chloride, propenoyl chloride, acryloylchloride, chlorid kyseliny akrylove, unii-8k23o56tg5, chlorid kyseliny akrylove czech, acrylylchloride
PubChem CID 13140
IUPAC Name prop-2-enoyl chloride
SMILES ClC(=O)C=C

Specifications

Infrared Spectrum Authentic
Odor Pungent
Refractive Index 1.4335 to 1.4350 (20°C, 589nm)
Linear Formula H2C=CHCOCl
Quantity 1 L
Beilstein 02, 400
Stabilizer 0.1% phenothiazine
Formula Weight 90.51
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Acryloyl chloride, 98%
Show More Show Less
Hazard Category LACHRYMATOR
Hazard Statement MOISTURE SENSITIVE
EINECSNumber 212-399-0
RTECSNumber AT7350000
TSCA TSCA
Recommended Storage Rechecking the purity prior to using this product is strongly recommended, because of its very short shelflife
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.