Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ACT 335827
GREENER_CHOICE

Catalog No. 498310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-831-0 10 mg
49-835-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-831-0 Supplier Tocris Bioscience™ Supplier No. 4983/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective OX1 antagonist

Potent and selective orexin OX1 receptor antagonist (Kb values are 41 and 560 nM for OX1 and OX2 receptors respectively). Elicits anxiolytic effects in vivo. Brain penetrant and orally available.

Chemical Identifiers

CAS 1354039-86-3
Molecular Formula C31H38N2O5

Specifications

Quantity 10 mg
Formula Weight 518.64
Molecular Weight (g/mol) 518.654
Chemical Name or Material ACT 335827
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.