Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACT1 Monoclonal Antibody (9ACT12), eBioscience™
GREENER_CHOICE

Catalog No. 5013029
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-130-29 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-130-29 Supplier Invitrogen™ Supplier No. 14404082
  • $388.00
    $291.65 / Each of 1

    Save $96.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The monoclonal antibody 9ACT12 reacts with human Act1 (NF-kappaB activator 1), also known as CIKS (a connection to IkB kinase and stress-activated kinase). Originally cloned for its ability to activate NF-kappaB and its direct interaction with IKKgamma, Act1 has been subsequently shown to play several roles in immunity. In B cells, it is a negative regulator of CD40 and BAFFR signaling affecting reduced survival. This regulation is through direct interaction of Act1 with CD40 or BAFFR and TRAF3. In addition, Act1 is involved in IL-17R signaling. It interacts with IL-17RA through the homotypic SEFIR interaction domain and is critical for downstream NF-kappaB- and C/EBP-mediated inflammatory responses. Applications Reported: This 9ACT12 antibody has been reported for use in immunoblotting (WB). Applications Tested: This 9ACT12 antibody has been tested by immunoblot of human peripheral blood cell lysates. This can be used at less than or equal to 1-5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

This gene encodes a protein involved in regulating responses to cytokines by members of the Rel/NF-kappaB transcription factor family. These factors play a central role in innate immunity in response to pathogens, inflammatory signals and stress. This gene product interacts with TRAF proteins (tumor necrosis factor receptor-associated factors) and either I-kappaB kinase or MAP kinase to activate either NF-kappaB or Jun kinase. Two alternative transcripts encoding different proteins have been identified. A third transcript, which does not encode a protein and is transcribed in the opposite orientation, has been identified. Overexpression of this transcript has been shown to reduce expression of at least one of the protein encoding transcripts, suggesting it has a regulatory role in the expression of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACT1
Applications Western Blot
Classification Monoclonal
Clone 9ACT12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TRAF3IP2
Gene Accession No. O43734
Gene Alias ACT1; adapter protein CIKS; AI429613; C6orf2; C6orf4; C6orf5; C6orf6; CANDF8; CIKS; connection to IKK and SAPK/JNK; NFkB-activating protein ACT1; nuclear factor NF-kappa-B activator 1; PSORS13; RGD1304768; RP3-487J7.3; TRAF3 interacting protein 2; TRAF3-interacting protein 2; TRAF3IP2
Gene Symbols TRAF3IP2
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10758
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.