Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam ACTH (1-24) (Human), ACTH/MC2 Receptor Agonist, Solid, 500 μg

Catalog No. 502706267
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6267 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6267 Supplier Abcam Supplier No. AB142251500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent ACTH/MC2 receptor agonist.

  • Molecular Weight: 2,933.4 Da
  • Function: ACTH/MC2 receptor agonist
  • Form: Solid
  • Constituents: 100% Adrenocorticotropic Hormone human
  • Storage Temperature: Store at -20°C
  • Shipping Temperature: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule ACTH (1-24) (Human), MC2 Receptor
Substrate Properties Peptide-based Substrate
Shipping Condition Ambient
Reactivity Human
Content And Storage Store at -20°C.
Form Solid
Quantity 500 μg
For Use With (Application) ACTH/MC2 receptor agonism
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.