Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam ACTH (1-24) Human, MC2 Receptor Agonist, Solid, 1 mg

Catalog No. 502706268
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6268 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6268 Supplier Abcam Supplier No. AB1422511MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly validated ACTH/MC2 receptor agonist for research applications.

  • Molecular Weight: 2933.4 Da
  • Form: Solid
  • Activity: ACTH/MC2 receptor agonist
  • Constituents: 100% Adrenocorticotropic Hormone human
  • CAS Number: 16960-16-0
  • Storage Temperature: Store at -20°C
  • Shipping Temperature: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Form Solid
Content And Storage Store at -20°C.
Shipping Condition Ambient
Target Molecule Adrenocorticotropic Hormone (Human)
Substrate Properties Peptide-based Substrate
Quantity 1 mg
For Use With (Application) ACTH/MC2 receptor agonist
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.