Learn More
Abcam ACTH (1-39) (human), endogenous melanocortin receptor 2 (MC2) agonist, 1MG
Supplier:  Abcam AB1411511MG
Encompass_Preferred
MW 4541 Da, Purity >95%. ACTH (1-39) (human), endogenous melanocortin receptor 2 (MC2) agonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6848