Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTH Recombinant Rabbit Monoclonal Antibody (9H5L7)

Catalog No. PI701293 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI701293 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI701293 Supplier Invitrogen™ Supplier No. 701293
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with non-human primate, human, porcine and rabbit based on sequence homology. Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

ATCH (adrenocorticotropic hormone) is a hormone which plays a major role in stimulating the adrenal cortex. It is formed through cleavage of the polypeptide precursor proopiomelanocortin (POMC), which also results in several other cleavage products including MSH, ACTH, and beta endorphin. ATCH is secreted from the anterior pituitary in response to the corticotropin-releasing hormone from the hypothalamus. It stimulates the secretion of glucocorticoids like cortisol, but has little control over the stimulation of mineralocorticoids like aldosterone, which is another major hormone of the adrenal cortex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTH
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 9H5L7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene Pomc
Gene Accession No. P01189, P01193, P01194
Gene Alias ACTH; adrenal corticotropic hormone; adrenocorticotropic hormone; adrenocorticotropin; alpha-melanocyte stimulating hormone; alpha-melanocyte-stimulating hormone; alphaMSH; alpha-MSH; BE; beta-endorphin; Beta-LPH; beta-melanocyte-stimulating hormone; beta-MSH; Clip; Corticotropin; Corticotropin-like intermediary peptide; corticotropin-lipotropin; Gamma-LPH; gamma-MSH; Lipotropin beta; lipotropin gamma; LPH; Melanocyte-stimulating hormone alpha; Melanocyte-stimulating hormone beta; Melanotropin alpha; melanotropin beta; Melanotropin gamma; met-enkephalin; MSH; NPP; opiomelanocortin prepropeptide; OTTHUMP00000119991; OTTHUMP00000200964; POC; Pomc; Pomc1; Pomc-1; Pomc2; Potential peptide; Precursor of MSH; pro-ACTH-endorphin; proopiomelanocortin; pro-opiomelanocortin; proopiomelanocortin (adrenocorticotropin/ beta-lipotropin/ alpha-melanocyte stimulating hormone/ beta-melanocyte stimulating hormone/ beta-endorphin); proopiomelanocortin preproprotein; proopiomelanocortin, beta (endorphin, beta); pro-opiomelanocortin-alpha; proopoimelanocortin, beta (endorphin, beta)
Gene Symbols Pomc
Host Species Rabbit
Immunogen Peptide corresponding to amino acids 147-172 of human ACTH.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18976, 24664, 5443
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.