Learn More
Diagnostic Biosystems ACTIN MUSCLE Actin, Muscle Specific
Supplier:  Diagnostic Biosystems PDM002
Encompass_Preferred
This antibody is specific to α and γ specific actin isomers from skeletal, cardiac and smooth muscle but does not recognize β and nonsmooth muscle γ actin isomers.
Catalog No. 50-238-5852
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.