Learn More
Cytoskeleton, Inc. Actin Protein ( >99% Pure): Human Platelet | 2x250ug
Supplier:  Cytoskeleton, Inc. APHL99A
Encompass
Non-muscle actin has been purified from human platelets. Each unit of platelets used in the preparation of non-muscle actin has been found to be non-reactive by an FDA approved test for HBsAg, HBcAb, HIV-1/2 ab, HIV-1 RNA, HTLV I/II ab, HCV ab, HCV RNA, and syphilis. Each unit of platelets has been ALT tested with results less than an established cutoff. The isotype composition of non-muscle actin is 85% β-actin and 15% γ-actin. Non-muscle actin has an approximate molecular weight of 43 kDa. The product is provided as a lyophilized white powder. The lyophilized protein is stable for 6 months when stored desiccated to <10% humidity at 4°C. The protein should be reconstituted to 10 mg/ml with distilled water. It will then be in the following buffer: 5 mM Tris-HCl pH 8.0, 0.2 mM CaCl2, 0.2 mM ATP, 5% sucrose, and 1% dextran.
Catalog No. NC1876244
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.