Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Actin (Rabbit)

Catalog No. 89932439 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-439 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-932-439 Supplier Abnova™ Supplier No. P5204
Add to Cart
Add to Cart

Used for SDS-PAGE

#REF!

Specifications

Accession Number P68135.1
For Use With (Application) SDS-PAGE
Molecular Weight (g/mol) 42kDa
Name Actin (Rabbit)
Preparation Method Native protein purified from Rabbit Muscle by modification of the procedure of Spudich and Watt, J. Biol. Chem., 246, 4866 (1971)
Quantity 1 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name ACTIN
Assays Binding studies; Endogenous internal controls to normalize gene expression studies.
Species Rabbit
Protein Tag None
Expression System Native protein purified from Rabbit Muscle by modification of the procedure of Spudich and Watt, J. Biol. Chem., 246, 4866 (1971)
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.