Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Actin Recombinant Rabbit Monoclonal Antibody (JJ09-29)
GREENER_CHOICE

Catalog No. PIMA532479 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532479 Supplier Invitrogen™ Supplier No. MA532479
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. In vertebrates there are three groups of actin isoforms: alpha, beta and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exists in most cell types as components of the cytoskeleton and as mediators of internal cell motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone JJ09-29
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene ACTB
Gene Accession No. P60709, P60710, P60711, P62736, P62737, P62738, P63259, P63260, P63261, P63267, P63268, P63269, P68032, P68033, P68035, P68133, P68134, P68136
Gene Alias 0610041G09Rik; 5C actin; A; A4V404_DROME; AA959943; a-actin; AAT6; Ac5C; act; act 5C; ACT1_DROME; ACT2; Act4; ACT-4; Act5; ACT-5; Act5c; Act-5C; Act5C-PA; Act5C-PB; Act5C-PC; Act5C-PD; Act5C-PE; acta; ACTA1; ACTA1 protein; acta1.S; acta2; Acta-2; ACTA3; ACTB; actb.L; ACTC; ACTC1; Actc-1; ACTE; Actg; ACTG1; Actg2; ACTGE; Actin; actin 2; actin 5 C; actin 5c; actin 7; actin A1; actin alpha 1; actin alpha 1, skeletal muscle; actin alpha 2, smooth muscle; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin beta; actin gamma 1; actin gamma 2, smooth muscle; actin homolog; actin, alpha 1, skeletal muscle; actin, alpha 1, skeletal muscle S homeolog; actin, alpha 2, smooth muscle, aorta; actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; Actin, alpha skeletal muscle; actin, alpha skeletal muscle S homeolog; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actin, beta; actin, beta L homeolog; actin, beta, cytoplasmic; actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; Actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; Actin, cytoplasmic type 5; actin, cytoplasmic, type 5; actin, gamma 1; actin, gamma 1 propeptide; actin, gamma 2; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; actin, gamma-enteric smooth muscle; Actin, gamma-enteric smooth muscle, intermediate form; Actin/BAP47; Actin5C; Actin-5C; actin-like protein; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; Actvs; Actx; AL023024; alpha 1 skeletal muscle actin; alpha actin; alpha actin 1; alpha actin 2; alpha-actin; alpha-actin cardiac; alpha-actin-1; Alpha-actin-2; alpha-actin-3; alphac-actin; Alpha-cardiac actin; alphaSMA; alpha-sma; alpha-smooth muscle actin; anon-EST:fe2D2; anon-fast-evolving-2D2; ArpA; ASD5; asma; a-SMA; AtACT7; Atcb; Bact; Bact; actin; B-actin; Bap47; beta actin; beta cytoskeletal actin; beta-actin; beta-actin FE-3; beta-actin/Bap47; BRWS1; BRWS2; cardiac actin; cardiac muscle alpha actin 1; cell growth-inhibiting gene 46 protein; cellular cytoskeletal beta-actin; CFTD; CFTD1; CFTDM; CG4027; CG4027-PA; CG4027-PB; CG4027-PC; CG4027-PD; CG4027-PE; chrX:5748184..5748304; CMD1R; CMH11; cyt5C; cytoplasmic 1; cytoplasmic 2; cytoplasmic actin; cytoplasmic beta-actin; cytoplasmic type 5; cytoskeletal beta actin; cytoskeletal gamma-actin; cytoskeletal protein; deafness, autosomal dominant 20; deafness, autosomal dominant 26; DEFORMED ROOT HAIRS 1; DER1; DFNA20; DFNA26; Dm-actin5C; Dmel\CG4027; Dmel_CG4027; E430023M04Rik; E51; ENHANCER OF LRX1 2; ENL2; epididymis luminal protein 176; F-actin; FIZ2; FRIZZY AND KINKED SHOOTS 2; G-actin; gamma 1 propeptide; gamma non-muscle actin; gamma-2-actin; Gamma-actin; gamma-enteric smooth muscle actin; GIG46; HEL-176; hypothetical protein LOC515610; I79_002310; I79_013242; I79_019066; l(1)G0009; l(1)G0010; l(1)G0025; l(1)G0079; l(1)G0117; l(1)G0177; l(1)G0245; l(1)G0330; l(1)G0420; l(1)G0486; LIGHT STRESS-REGULATED 2; LOW QUALITY PROTEIN: actin, alpha skeletal muscle; LOW QUALITY PROTEIN: actin, aortic smooth muscle; LSR2; LVNC4; M32055; mpfd; MYH9.2; MYH9_2; MYMY5; NEM1; NEM2; NEM3; nemaline myopathy type 3; pan actin; PS1TP5-binding protein 1; PS1TP5BP1; RE02927p; RP5-1068B5.2; SHPM; similar to b-actin; similar to beta actin; skeletal alpha actin; sma; SMalphaA; SMGA; smooth muscle alpha-actin; smooth muscle gamma-actin; T11; type 5; unnamed protein product; vascular smooth muscle alpha-actin; VSCM; wu:fb63d03; XELAEV_18029136mg; XELAEV_18045052mg
Gene Symbols Acta1, ACTA2, ACTB, ACTC1, ACTG1, ACTG2
Host Species Rabbit
Immunogen Synthetic peptide within Human Actin aa 45-80.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11459, 11461, 11464, 11465, 11468, 11475, 25365, 287876, 29275, 29437, 322509, 58, 59, 60, 70, 71, 72, 81633, 81822
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.