Learn More
ATCC Actinobacillus pleuropneumoniae; 4074
Supplier:  ATCC 27088
Encompass
Bacterial type strain isolated from the lung of swine. This strain is genome sequenced.
Catalog No. NC2302887
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.