Learn More
ATCC Actinobacillus pleuropneumoniae; serotype 3
Supplier:  ATCC 27090
Encompass
Actinobacillus pleuropneumoniae strain S 1421 was isolated in Switzerland from a swine periarticular abscess. This bacterial culture can be used in susceptibility testing and respiratory research.
Catalog No. NC2293621
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.