Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Actinomycin D Inhibitor
SDP

Catalog No. NB1229 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1229 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1229 Supplier Novus Biologicals™ Supplier No. NB1229

missing translation for 'validMainframeMsg'

Low molecular weight, organic compound that may help regulate biological process.

Biological Activity: Anti-neoplastic antibiotic. Inhibits RNA polymerase, potent inducer of apoptosis
Storage: Store at 4°C.

Note: Inhibitors are guranteed for 1 year from date of receipt.

Specifications

Color Red
Product Type Inhibitor
Target Proteins Actinomycin D
CAS 50-76-0
Molecular Formula C62H86N12O16
Formula Weight 1255.44g/mol
Solubility Soluble to 50mM in DMSO
Content And Storage Store at 4°C
For Use With (Application) Inhibits apoptosis
Quantity 10 mg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.