Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ReadyProbes™ Reagent F-Actin Phalloidin Conjugates

Catalog No. R37112
Encompass_Preferred
Change view
Click to view available options
Color:
Green
Red-Orange
Excitation Wavelength Range:
495/518
540/565
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Excitation Wavelength Range
R37112 Red-Orange 540/565
R37110 Green 495/518
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R37112 Supplier Invitrogen™ Supplier No. R37112
Only null left
Add to Cart
Add to Cart

Includes

2 x 2.5 mL dropper bottles

Perfect for fixed cells and tissue, ReadyProbes Reagent F-actin phalloidin conjugates offer clear visualization of actin filaments, making it perfect for cell biology, neuroscience, cancer research, and more.

ActinGreen 488 ReadyProbes Reagent is a selective, high-affinity F-actin probe conjugated to our bright, photostable, green-fluorescent Alexa Fluor 488 dye. ActinGreen 488 reagent is a room temperature-stable solution of Alexa Fluor 488 Phalloidin (Cat. No. A12379) packaged in a dropper bottle. ActinRed 555 ReadyProbes Reagent is a comparable F-actin stain conjugated to the red-orange fluorescent dye tetramethylrhodamine (TRITC). ActinRed 555 reagent is a room temperature-stable solution of Rhodamine Phalloidin (Cat. No. R415) packaged in a dropper bottle.

These ready-to-use solutions combines superior brightness with exceptional F-actin staining. Phalloidin is a bi-cyclic peptide, commonly used in imaging applications to selectively label F-actin. Fluorescently labeled phalloidin has several advantages over antibodies for actin labeling, including virtually identical binding properties with actin from different species of plants and animals and low non-specific binding.

Features

  • High-affinity staining of F-actin with superior specificity compared to antibody methods
  • ActinGreen 488 Excitation/Emission: (495/518 nm), detected through standard GFP and FITC filter sets
  • ActinRed 555 Excitation/Emission: (540/565 nm), detected through standard RFP and TRITC filters sets
  • Use counterstain (i.e., DAPI or NucBlue ReadyProbes reagent) as desired

ReadyProbes format advantages

  • Ready-to-use formulation—no need to dilute, weigh, or pipette
  • Just use two drops per mL of medium to stain, incubate 30 minutes, wash, then image
  • Stable at room temperature—keep handy on the benchtop

Applications

  • For fixed and permeabilized cells or tissue
  • Fluorescence microscopy (IF/IHC/ICC)
  • High content screening, high content imaging
  • Flow cytometry

Suggestions for use

  • Both ActinRed 555 and ActinGreen 488 reagents may be added directly to fixed cells in full medium or buffer solutions
  • In most cases 2 drops/mL and an incubation time of 15–30 minutes is sufficient for bright actin staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained.
TRUSTED_SUSTAINABILITY

Specifications

Color Red-Orange
Content And Storage

2 x 2.5 mL dropper bottles

Store at ≤ 25°C.

Excitation Wavelength Range 540/565
Dye Type TRITC (Tetramethylrhodamine Isothiocyanate)
For Use With (Equipment) Confocal Microscope, EVOS™, Floid™ Cell Imaging System, Fluorescence Microscope, Flow Cytometer, Floid™ Cell Imaging System
Quantity 2 dropper bottle(s) kit
Detection Method Fluorescence
Description ActinRed™ 555 ReadyProbes™ Reagent (Rhodamine phalloidin)
Emission Visible
Form Liquid
Product Line ActinRed, ReadyProbes
Technique Fluorescence Intensity
Label Type Fluorescent Dye
Product Type Phalloidin
SubCellular Localization Cytoskeleton
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.