Learn More
Superior Glove™ Action™ Aramid Fiber Knit Leather Gloves
Catalog No. 17489653
Change view
Click to view available options
Size:
Small
Medium
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-489-653 Small
17-489-654 Medium
17-489-656 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-489-653 Supplier Superior Glove™ Supplier No. SKGLPLC/S
This item is not returnable. View return policy
Gloves are constructed from aramid fibers for cut protection, with full-grain leather palms for oil/water resistance.
Specifications
Construction Cut & Sewn
Color Natural, Yellow
Cut Resistance Rating ANSI A2
Hand Specific
Material Side Split Cowhide Leather
Product Type General Purpose Gloves
Size Small
Glove Cuff Color Gray
Glove Cuff Style Gauntlet
Series SKGLPLC
Length (English) 4 in.
Length (Metric) 10.2 cm