Learn More
Superior Glove™ Action™ Aramid Fiber String Knit Cut-resistant Gloves, Grain-Leather Palm
Description
- Leather palms wrap around sides of fingers, while thumb is all leather
- Glove body made of knit Kevlar™ aramid fiber for cut and slash resistance, plus flame-resistance
- Palms attached to knit shell using Kevlar™ aramid fiber thread for greater seam integrity
Specifications
Specifications
| Cut Resistance Rating | ANSI A2 |
| Material | Aramid Fiber, Leather |
| Product Type | Cut-resistant Gloves |
| Size | Medium |
| For Use With (Application) | sheet Metal, Metal stamping, Glass handling |
| Glove Cuff Style | Knit |