Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Action™ Aramid Fiber String Knit Cut-Resistant Gloves

Catalog No. 17400101
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-400-101 Small
17-400-100 X-Small
17-400-102 Medium
17-400-103 Large
17-400-104 X-Large
17-400-105 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-400-101 Supplier Superior Glove™ Supplier No. SKLP/S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Aramid fiber cut and slash-resistant gloves feature leather palm and thumb to offer protection and extended wear.

  • 7-gauge
  • Leather palms wrap around sides of fingers
  • Leather thumb
  • Durable tri-tan leather offers greater protection, longer wear
  • Shell made of aramid fiber for cut and slash resistance
  • Flame-resistance qualities
  • Palms attached to knit shell using sewing thread made with Kevlar™ aramid fiber for added seam integrity

Specifications

Color Yellow, Gray
Cut Resistance Rating ANSI A2
Material Aramid Fiber, Split Leather
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) sheet Metal, Metal stamping, Glass handling
Glove Cuff Style Knit
Model SKLP/S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.