Learn More
New Pig Action Pump Stainless Steel Piston Hand Drum Pump
SDP
Catalog No. 17987830 Shop All New Pig Corporation Products
Catalog No. 17-987-830 Supplier New Pig Supplier No. DRM343
Add to Cart
Add to Cart
Quickly dispense flammable fluids from your drums with this versatile stainless steel piston operated pump
  • Safely dispenses flammable liquids: The pump is designed to be used with flammable fluids, and its earthing stud allows it to be grounded for static discharge, enhancing safety.
  • Durable stainless steel construction: Made of 316 stainless steel and PTFE, the pump is highly resistant to a wide range of chemicals and solvents, ensuring longevity and reliable performance.
  • High-performance piston action: The piston-operated design allows you to quickly and efficiently dispense up to 22 oz. of fluid per stroke.
  • Ergonomic and easy to use: A comfortable T-handle design and smooth piston action make the pump easy to operate, reducing user fatigue.
  • Versatile application: This pump fits standard 15, 30, and 55 ga. drums with 2 in. NPT bungs and is compatible with a variety of fluids including gasoline, diesel fuel, kerosene, and light oils.
  • Corrosion-resistant and reliable: Its robust construction makes it an ideal choice for dispensing corrosive liquids in demanding industrial environments.