Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 43-216 Cowhide Leather Industrial Heat-Resistant Gloves

Catalog No. 19010150 Shop All Ansell Products
Change view
Click to view available options
Size:
9
11
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-150 11
19-010-148 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-010-150 Supplier Ansell Supplier No. 43216110
  • $1,411.00
    $1,056.65 / Case of 36

    Save $354.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Premium split cowhide leather combined with Aramid Fiber welted seams ensure durability in use, while also providing protection during high temperature applications.

  • Premium split cowhide leather and welted seams with Aramid Fiber thread
  • Improved 3-dimensional thumb design provides fine dexterity and control
  • Cotton liner enhances comfort and thermal insulation
  • Long cuff extends protection over the forearm
  • Suitable for stick and MIG welding, as well as torch cutting and grinding
  • Gloves are not liquid proof and should not to be used with chemicals
  • Do not use in heavy-duty cut applications
  • Keep away from sources of ozone or ignition
  • Store in a cool and dry place, away from direct sunlight
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Coating Material Uncoated
Color Yellow
Latex Free Yes
Material Cowhide Leather, Aramid Fiber
Product Type Welding Gloves
Size 11
For Use With (Application) Automotive, Machinery/Equipment, Metal Fabrication, and MIG/Stick Welding
Glove Cuff Style Gauntlet
Liner Material Cotton
Series ActivArmr™ 43-216
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.