Learn More
Description
- Engineered for rugged and demanding jobs
- Patented Intercept Technology reinforced with stainless steel and Kevlar™ aramid fibers
- Protection from punctures, cuts and abrasions (Cut Level 4, Abrasion Level 4)
- Reinforced ridges offer additional palm and knuckle protection
- Foam nitrile coating provides enhanced wet or dry grip
- “Second skin” feel gives dexterity and precision all day long
- Adjustable wrist closure with pull-tab and ID nameplate
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 4 |
| Certifications/Compliance (Europe) | CE, EN 420:2003 + A1:2009; Category II |
| Construction | Knit |
| Coating Material | Nitrile Foam |
| Color | Black, Gray, Orange |
| Cut Resistance Rating | ANSI A4 |
| Product Type | Cut-resistant Gloves |
| Size | X-Large |
| EN388 Standard | EN 388:4X32D, EN 388:4432 |
| For Use With (Application) | Demolition, handling sheet metal, masonry, woodwork, digging, general construction, heavy cutting/ sawing, steel work (tying) |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
