Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 97-681 PVC-Coated Cold Resistant Waterproof Gloves

Catalog No. 19500970 Shop All Ansell Products
Change view
Click to view available options
Size:
8
9
10
11
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-970 11
19-500-968 9
19-500-969 10
19-500-967 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-500-970 Supplier Ansell Supplier No. 97681110
  • $276.00
    $224.65 / Pack of 6

    Save $51.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,455.00
    $1,415.65 / Case of 6 PK

    Save $39.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An insulating acrylic liner combined with RIPEL™ Technology helps to keep hands dry and warm for an extended period of time. PVC coating enhances grip capabilities and remains flexible, even in the coldest conditions.

  • PVC foam coating provides grip and flexibility in temperatures down to -25°C (-15°F)
  • RIPEL™ Technology keeps liquids out to keep hands dry and warm
  • Two-layer design protects agains liquid and mechanical risks, while also providing warmth and comfort
  • Thin enough to handle small objects, but protective enough for harsh environments
  • Helps to prevent frostbite
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Coating Material PVC
Color Blue
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Cold-resistant Gloves
Size 11
EN388 Standard 2231B
For Use With (Application) Food Processing, Infrastructure Maintenance, and Oil/Gas
Glove Cuff Style Gauntlet
Liner Material Acrylic
Series ActivArmr™ 976-81
Temperature (English) Min -15°F
Temperature (Metric) Min. -25°C
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.