Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- 15 gauge seamless HPPE Knit Glove keep hands cool and comfortable
- PU dip provides excellent grip and dexterity
- HIVIZ™ design increases awareness for safety.
- Dirt-masking dark liner is well-suited for slightly oily or dirty environments helping improve wear life
- Designed for dynamic environments which have cut and impact hazards as well as the need for precision
- Thoughtfully designed In Tact Bumpers maximize protection, flexibility, and dexterity
Specifications
Specifications
| Certifications/Compliance | ANSI Level 4 Abrasion and ANSI A2 Cut Protection. Accordance with EN 388:2016. |
| Coating Material | Polyurethane |
| Color | Gray, Orange |
| Cut Resistance Rating | ANSI A2 |
| Hand | Specific |
| Latex Free | No |
| Material | HPPE, Lycra, Nylon |
| Product Type | Cut-resistant Gloves |
| Size | 6 |
| For Use With (Application) | Heavy Equipment Servicing, Light and Heavy Assembly, Maintenance, Production Services/Processing, Shop Work, Transportation and Logistics |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?