Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 97-631 Cold-Resistant Two-Layer Gloves

Catalog No. 19500762 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-762 7
19-500-763 8
19-500-764 9
19-500-765 10
19-500-766 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-500-762 Supplier Ansell Supplier No. 97631070
Add to Cart
Add to Cart

Two-Layer gloves feature a unique combination of warmth, dexterity, and flexibility, allowing the user feel everything but the cold.

  • PVC coating provides excellent grip and flexibility, even in the coldest temperatures
  • Flexible nylon exterior protects against mechanical risks, including cut, and contours to the hand permitting maximum dexterity
  • Soft and insulative acrylic interior provides comfort and warmth
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category ll; REACH Compliant; EN51112X, EN 388:2231B
Coating Material PVC
Color Black
Cut Resistance Rating ANSI A2
Material Acrylic, Nylon, PVC
Size 7
For Use With (Application) Construction, outdoor winter use, cold storage, utilities
Glove Cuff Style Knit
Liner Material Acrylic, Nylon
Length (English) 9.8 to 11.2 in.
Length (Metric) 25 to 28.5 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.