Learn More
Activated Mac-1 (CD11b) Mouse anti-Human, Brilliant Violet 421, Clone: CBRM1/5, BD
Shop All BD Biosciences ProductsDescription
The CBRM1/5 monoclonal antibody specifically recognizes an activated form of the CD11b molecule that is expressed on activated monocytes, granulocytes, NK cells, and subsets of T and B cells. CD11b is also known as the alpha chain of either Mac-1 (MAC-1A/Mac-1 ) or Complement receptor 3 (CR3A). CD11b is a ñ165 kDa type I transmembrane glycoprotein that is encoded by ITGAM (Integrin alpha M). CD11b associates with CD18 (Integrin beta 2) to form the CD11b/CD18 complex, which is also known as the Mac-1 integrin or the Complement receptor 3 (CR3). CD11b functions in cell-cell and cell-substrate interactions involved in leucocyte activation, adhesion, migration, phagocytosis, and other effector functions. CD11b/CD18 serves as a receptor for multiple ligands including iC3b, CD50 (ICAM-3), CD54 (ICAM-1), CD102 (ICAM-2), and fibrinogen. The CBRM1/5 reportedly inhibits Mac-1-dependent adhesion of stimulated neutrophils to CD54 or fibrinogen.
Specifications
Specifications
| Antigen | Activated Mac-1 (CD11b) |
| Classification | Monoclonal |
| Clone | CBRM1/5 |
| Conjugate | Brilliant Violet 421 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | MAC-1A, Mac-1, ITGAM, Integrin alpha M, CR3A, CR-3 alpha, Mo1, SLEB6 |
| Host Species | Mouse |
| Immunogen | Human Mac-1 Protein |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.