Learn More
CPC Scientific H-Tyr-Gly-Val-Tyr-Thr-Lys-Val-Ser-Arg-Tyr-Leu-Asp-Trp-Ile-His-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific ACP002A
Encompass
Activated protein C (APC), proteolyticly inactivate the coagulation cofactors Va and VIIIa. Human APC residues 390-404 are essential for its anticoagulant activity.ONE-LETTER SEQUENCE: YGVYTKVSRYLDWIHMOLECULAR FORMULA: C91H130N22O23MOLECULAR WEIGHT:1900.17STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [146340-20-7], RESEARCH AREA: CardiovascularREFERENCES: R.M.Mesters et al., J. Biol. Chem., 266, 24514 (1991)
Catalog No. 50-210-8590
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.