Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activator Reagent, Solstice

Catalog No. 60090105 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
56 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-105 200 L
60-090-111 56 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-090-105 Supplier Solstice Supplier No. BC721200
Add to Cart
Add to Cart
Request a Quote

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.25M ETT in Acetonitrile

TRUSTED_SUSTAINABILITY

Specifications

CAS 89797-68-2, 75-05-8
CAS Max % 4.00%, 96.00%
Quantity 200 L
Flash Point 7°C
Packaging Returnable Steel Drum
Physical Form Liquid
Purity Grade Notes Activator reagents
Boiling Point 82°C.The physical data is that of the main component.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.