Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activator Solution, 0.25M 5-(Ethylthio)-1H-Tetrazole in Acetonitrile, Novabiochem™, MilliporeSigma™

Catalog No. MB0152P4000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MB0152P4000 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB0152P4000 Supplier MilliporeSigma Supplier No. BI0152P4000
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-05-8
Molecular Formula CH3CH2S-CHN4 in CH3CN
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn

Specifications

Color Colorless
Boiling Point 81.6°C (1013 hPa)(Acetonitrile)
Flash Point 7.2°C
Packaging Glass Bottle
Quantity 4 L
Vapor Pressure 97 hPa (20°C)
Physical Form Liquid
Chemical Name or Material Acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.