Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sqwincher™ Activity Drinks, Ready-to-Drink

Catalog No. 19003198A
Change view
Click to view available options
Flavor:
Cherry
Fruit Punch
Lemon Lime
Orange
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flavor
19-003-198A Orange
19-003-198C Fruit Punch
19-003-198D Lemon Lime
19-034-148 Cherry
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-003-198A Supplier Sqwincher™ Supplier No. 30534OR
Add to Cart
Add to Cart

Thirst-quenching drink that's low in sodium and high in potassium

  • Formulated with electrolytes and nutrients
  • Replenishes fluids, minerals, and sugars depleted through physical exertion
  • Prevents or reduces severity of heat-stress disorders
  • Provides supplemental source of energy
  • Available as ready-to-drink ActivityPack™ in handy cartons and in sports bottles

Specifications

Product Type Activity Drink
Flavor Orange
Format Sports Bottle
For Use With (Application) Provides supplemental source of energy, prevents or reduces severity of heat-stress disorders
Quantity 24 Cs.
Volume (English) 20 oz.
Volume (Metric) 591 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.