Learn More
MyBioSource Human ELISA Kits
SDP
Supplier:  MyBioSource MBS720075196STRIPWELL
Encompass_Preferred
The MyBioSource Human ELISA Kits are used for the quantitative measurement of human proteins, peptides, hormones, immunoglobulins, cytokines, and more in serum, plasma, cell culture supernates, and other biological fluids. They utilize a quantitative sandwich enzyme immunoassay technique and come with all necessary reagents to get reliable results in less than 3 hours. These ELISA kits have high sensitivity, excellent specificity, and no cross-reactivity with analogues. They have been validated and tested for consistent results with intra- and inter-assay precision. The kits are suitable for both manual and automated operation.
Catalog No. NC2410989
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.