Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACVR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674804
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-804 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-804 Supplier Invitrogen™ Supplier No. PA112641
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

ACVR1 Polyclonal Antibody for Western Blot

Activin A receptor, type I (ACVR1) belongs to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. ACVR1 signals a specific transcriptional response in concert with activin type II receptors. The ACVR1 gene encodes activin A type I receptor, which signals a particular transcriptional response in concert with activin type II receptors. Mutations in ACVR1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVR1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene ACVR1
Gene Accession No. Q90ZK6
Gene Alias activin A receptor type 1; activin A receptor type I; activin A receptor, type 1; activin A receptor, type I; activin A receptor, type II-like kinase 2; activin receptor I precursor; activin receptor type I; Activin receptor type-1; activin receptor-like kinase 2; activin type I receptor; ACTRI; ActR-I; ActRIA; Acvr; ACVR1; ACVR1A; ACVRLK2; ALK2; Alk-2; ALK2-B; Alk8; ATVR1; D330013D15Rik; FOP; hydroxyalkyl-protein kinase; serine/threonine-protein kinase receptor; serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; Tgfb1; Tsk7L; TSK-7L; TSRI; TSR-I; type I TGF B receptor
Gene Symbols ACVR1
Host Species Sheep
Immunogen Recombinant protein corresponding to the extracellular domain of chicken ALK2.
Purification Method Ammonium Sulfate Precipitation
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 395246
Target Species Chicken
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.