Learn More
MyBioSource ACVR2B antibody
SDP
Supplier:  MyBioSource MBS81136290.3 MG
Encompass_Preferred
The MyBioSource ACVR2B antibody is a rabbit polyclonal antibody raised against a partial recombinant ACVR2B of human origin. It is supplied as 100 μg purified antibody in 200 μl of PBS with 0.09% sodium azide and 50% glycerol. The immunogen is a recombinant protein of human ACVR2B. This antibody has been tested in WB, ELISA, and IHC applications. It is recommended for detection of ACVR2B of human origin by WB, ELISA, IHC, and other antibody based applications. The molecular weight of ACVR2B is predicted to be 60 kDa according to the sequence.
Catalog No. NC2884809