Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Human ACVRL1 (ALK1), GST Tag Recombinant Protein

Catalog No. PV4884
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4884 1 mg
PV4883 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV4884 Supplier Thermo Scientific™ Supplier No. PV4884

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.

Activin A receptor type II-like 1 (ACVRL1) is a type I cell-surface receptor for the TGF-beta superfamily of ligands. ACVRL1, similar to other type 1 receptors, has a conserved serine-threonine kinase subdomain, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. Mutations in ACVRL1 are associated with hemorrhagic telangiectasia type 2. Mutations in ACVRL1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P37023
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 150mM NaCl, 0.5mM EDTA, 0.02% polysorbate 20, 50% glycerol, 2mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 94
Molecular Weight (g/mol) 68.9 kDa
Name Human ACVRL1 (ALK1), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias activin A receptor like type 1; activin A receptor type II-like 1; activin A receptor type IL; activin A receptor, type II-like 1; activin A receptor, type II-like kinase 1; Activin receptor like kinase 1; activin receptor-like kinase 1; activin receptor-like kinase-1; Acvrl1; Acvrlk1; AI115505; AI427544; ALK1; ALK-1; HHT; HHT2; ORW2; R3; R-3; Serine/threonine-protein kinase receptor R3; SETHKIR; SKR3; TGF-B superfamily receptor type I; TSR-I
Common Name ACVRL1
Gene Symbol ACVRL1
Kinase Family STKR Kinase Family
Protein Length 139-503
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 139-503
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: TKL Group
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Catalytic
Form Liquid
Research Category Drug Development
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.