Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACVRL1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7205804 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9691 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9691 Supplier Proteintech Group Inc Supplier No. 147451AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The TGF-beta superfamily of ligands exerts its biological activity by binding to heteromeric receptor complexes two types (I and II) of the serine/threonine kinases. ALK1, like the other type I receptors, contains a cysteine rich domain with conserved cysteine spacing in the extracellular region and a glycine and serine rich domain preceding the kinase domain. The reduced human ALK1 monomer has a calculated molecular mass of approximately 37.3 kDa. Due to glycoslyation, the recombit human protein migrates as a 50-55 kDa protein in SDS-PAGE under reducing conditions. Human and mouse ALK1 share approximately 71% amino acid sequence identity in their extracellular domains. ALK1 is highly expressed in endothelial cells and other vascular tissues.

Specifications

Antigen ACVRL1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACVRL1
Gene Accession No. P37023
Gene Alias Activin receptor like kinase 1, ACVRL1, ACVRLK1, ALK 1, ALK1, HHT, HHT2, ORW2, SKR3, TSR I
Gene Symbols ACVRL1
Host Species Rabbit
Immunogen ACVRL1 Fusion Protein Ag6449
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.