Learn More
Selleck Chemical LLC ACY-738 2mg 1375465-91-0
SDP
Supplier:  Selleck Chemical LLC S86482MG
Encompass_Preferred
ACY-738 inhibits HDAC6 with low nanomolar potency (IC50=1.7 nM) and a selectivity of 60- to 1500-fold over class I HDACs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8138
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.