Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acyclovir, R&D Systems™
SDP

Catalog No. 251350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 25-135-0 Supplier R&D Systems Supplier No. 2513/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibits viral DNA polymerase; antiherpetic agent

Antiviral agent, active against herpes simplex viruses HSV-1 and HSV-2 (EC50 values are 0.85 and 0.86 μM respectively). Interferes with viral DNA polymerization through competitive inhibition with guanosine triphosphate. Induces apoptosis in cells transfected with HSV-TK (suicidal gene therapy).

Chemical Identifiers

CAS 59277-89-3
Molecular Formula C8H11N5O3
Molecular Weight (g/mol) 225.21
MDL Number MFCD00057880
InChI Key MKUXAQIIEYXACX-UHFFFAOYSA-N
Synonym acyclovir, aciclovir, acycloguanosine, zovirax, vipral, virorax, wellcome-248u, 9-2-hydroxyethoxy methyl guanine, zovir, aciclovirum
PubChem CID 2022
ChEBI CHEBI:2453
IUPAC Name 2-amino-9-[(2-hydroxyethoxy)methyl]-6,9-dihydro-1H-purin-6-one
SMILES NC1=NC2=C(N=CN2COCCO)C(=O)N1

Specifications

Quantity 50 mg
Formula Weight 225.21
Percent Purity >99%
Product Line Tocris
Chemical Name or Material Acyclovir
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.