Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACYP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120381 Supplier Invitrogen™ Supplier No. PA5120381
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: MCF7, Mouse brain, Mouse heart, Mouse kidney, Rat skeletal muscle. Immunogen sequence: MSTAQSLKSV DYEVFGRVQG VCFRMYTEDE ARKIGVVGWV KNTSKGTVTG QVQGPEDKVN SMKSWLSKVG SPSSRIDRTN FSNEKTISKL EYSNFSIRY.

Its physiological role is not yet clear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACYP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACYP2
Gene Accession No. P14621, P35745, P56375
Gene Alias 2310004B09Rik; acylphosphatase 2; acylphosphatase 2, muscle type; acylphosphatase, muscle type isozyme; Acylphosphatase-2; acylphosphate phosphohydrolase 2; ACYM; ACYP; ACYP2; testicular tissue protein Li 11
Gene Symbols ACYP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-99 of human ACYP2 (NP_612457.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364224, 75572, 98
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.