Learn More
Vector Biolabs The Adenovirus Co Cre Recombinase Adenovirus (Ad-CMV-iCre)
Supplier:  Vector Biolabs The Adenovirus Co 1045
Encompass_Preferred
This Cre recombinant adenovirus expresses Cre recombinase, a Type I topoisomerase from P1 bacteriophage that catalyzes site-specific recombination of DNA between loxP sites. loxP is a 34 bp DNA sequence at which confers directionality. Cre recombinase is used as a tool to genetically modify genes, such as to delete a segment of DNA flanked by LoxP sites in cultured cells or experimental animals.This adenovirus expresses a codon improved Cre (iCre) under a CMV promoter.Ad-Cre Video
Check out this instruction video on using our adenovirus Cre to knockout LoxP flanked gene in primary mouse embryonic fibroblasts (MEF).
Catalog No. NC9349494