Learn More
Vector Biolabs The Adenovirus Co AD-CMV-GFP
Supplier:  Vector Biolabs The Adenovirus Co 1060
Encompass_Preferred
Ad-CMV-GFP; Product Category: Human Adenovirus Type5 (dE1/E3); Green Fluorescent Protein; Titer: 10 10 PFU/mL; 200uL; Storage Buffer: DMEM w/2% BSA and 2.5% Glycerol; 1/PK
Catalog No. NC9349496
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.