Learn More
Vector Biolabs The Adenovirus Co Cre Recombinase Adenovirus (Ad-Cre-GFP)
Supplier:  Vector Biolabs The Adenovirus Co 1700
Encompass_Preferred
Cre Recombinase is a Type I topoisomerase from bacteriophage P1 that catalyzes the site-specific recombination of DNA between loxP sites. loxP is a 34 bp DNA sequence at which confers directionality. Cre recombinase is used as a tool to genetically modify genes, such as to delete a segment of DNA flanked by LoxP sites in cells or experimental animals.This adenovirus expresses both Cre recombinase and GFP as marker. Cre and GFP are driven by 2 separate promoters.Ad-Cre Video
Check out this instruction video on using our adenovirus Cre to knockout LoxP flanked gene in primary mouse embryonic fibroblasts (MEF).
Catalog No. NC1579991
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.