Learn More
Liofilchem™ AD Fosfomycin
SDP
Catalog No. 22778084 Shop All Liofilchem Products
Change view
Click to view available options
No. per Pack:
1 Ea.
6/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
22-778-084 1 Ea.
22-778-085 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.
Catalog No. 22-778-084 Supplier Liofilchem™ Supplier No. 77001
Only null left
Add to Cart
Add to Cart
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.
AD Fosfomycin is a 12-well panel containing the antibiotic incorporated into an agar medium in different concentrations. It is used to perform the AD method for the antimicrobial susceptibility testing of fosfomycin as recommended by CLSI and EUCAST standards.
  • 1 panel of AD Fosfomycin 0.25-256
  • 2 Reading Templates (transparent and black backgrounds)
  • 1 Instructions Sheet also available at www.liofilchem.com/ifu-sds
Specifications
Antimicrobial Agents Fosfomycin
Concentration 0.25 to 256 μg/mL
Product Type Panel
Sterility Non-sterile
Test Type Media Testing
No. per Pack 1 Ea.