Learn More
Description
- 12-well panel containing the antibiotic incorporated into an agar medium in different concentrations
- i.e. 11 two-fold dilutions (0.032 to 32 μg/mL)
- Used to perform AD (Agar Dilution) method
- Not for use directly with clinical or other specimens
- Microorganism to be tested must be pure culture on a dermatophyte culture medium supplemented with cycloheximide and chloramphenicol
- For mixed culture, selected colonies should be purified by subculturing
- Results are read visually at the end of incubation
Specifications
Specifications
| Form | Agar |
| Format | Dilution Panel |
| Manufacturing Origin | Italy |
| Product Type | Agar Medium |
| Sterility | Non-sterile |
| Target Organism Class | Enterobacterales |
| Test Type | Antibiotic Susceptibility Sensitivity |
| Quantity | 1 Ea. |
| Species | Escherichia coli |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
