Learn More
Vector Biolabs The Adenovirus Co AD(RGD)-CMV-GFP
Supplier:  Vector Biolabs The Adenovirus Co 1768
Encompass_Preferred
Ad infection is initiated by recognition of the native Ad5 receptor, coxsackievirus-adenovirus receptor (CAR), on target cells by the carboxy-terminal portion (i.e., knob) of the fiber protein. Therefore the infection efficiency of adenovirus is very low for cells with low or no CAR receptors. Arg-Gly-Asp (RGD) motif was introduced in the surface-exposed loops of adenovirus fiber knob, this enables the adenovirus to bypass CAR and mediate cell entry via RGD binding integrins. So the RDG modified adenovirus can be used for "retargeting" cells that regular adenovirus doesn't work well. The infection effeciency of RDG modified adenovirus can be tens or hundreds of times higher than standard Ad5 in some cells, including human or mouse macrophages, T cells, synoviocytes, islet grafts, adipocytes, MEF, etc.This is a RGD modified replication-deficient adenovirus expressing eGFP under a CMV promoter.
Catalog No. NC1905900
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.