Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM10 Mouse anti-Human, Unconjugated, Clone: 11G2, MilliporeSigma™

Catalog No. p-7181778 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN23172MI 25 μL
MABN23171MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN23172MI Supplier MilliporeSigma Supplier No. MABN231725UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ADAM10 Monoclonal antibody specifically detects ADAM10 in Human samples. It is validated for Electron Microscopy, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM10
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 11G2
Conjugate Unconjugated
Gene Accession No. NP_001101.1
Gene Alias Disintegrin and metalloproteinase domain-containing protein 10;EC:3.4.24.81;CDw156;Kuzbanian protein homolog;Mammalian disintegrin-metalloprotease;CD156c
Gene Symbols ADAM10, KUZ, MADM
Host Species Mouse
Immunogen Mixture of HEL human erythroleukemia cells and Jurkat cells.
Molecular Weight of Antigen 84.14 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 11G2 is a mouse monoclonal antibody that specifically detects human Disintegrin and metalloproteinase domain-containing protein 10 (ADAM10).
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.