Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM10 Recombinant Rabbit Monoclonal Antibody (JM32-11)
GREENER_CHOICE

Catalog No. PIMA532616 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532616 Supplier Invitrogen™ Supplier No. MA532616
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Proinflammatory cytokine tumor necrosis factor-a (TNF-a) contributes to a variety of inflammatory responses and programmed cell death. Notch receptor and its ligand participate in cell fate decisions during vertebrate development and are associated with several human disorders, including a T-cell lymphoma. TNF-a, notch and its ligand delta are all membrane-bond molecules, which are cleaved by proteases to release mature proteins or functional receptor. ADAM10, a metalloprotease-disintegrin in the family of mammalian ADAM (for a disintegrin and metalloprotease), was recently identified to cleave TNF-a, notch and its ligand delta (1-3). The genes encoding human, mouse, and bovine ADAM10 were recently cloned and designated ADAM 10, kuzbanian (KUZ), and MADM, respectively. ADAM10 mRNA is expressed in a variety of human and bovine tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM10
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone JM32-11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% Glycerol and 0.2% ProClin 950; pH 7.4
Gene Adam10
Gene Accession No. O14672, O35598, Q10743
Gene Alias 1700031C13Rik; a disintegrin and metallopeptidase domain 10; a disintegrin and metalloprotease domain (ADAM) 10; a disintegrin and metalloprotease domain 10; a disintegrin and metalloproteinase; AD10; AD18; ADAM; ADAM 10; ADAM metallopeptidase domain 10; ADAM10; ADAMs; CD156c; CDw156; Disintegrin and metalloproteinase domain-containing protein 10; HsT18717; KUZ; kuzbanian; kuzbanian protein homolog; MADM; Mammalian disintegrin-metalloprotease; metalloendopeptidases; metalloproteinase; myelin-associated metalloproteinase; RAK
Gene Symbols Adam10
Host Species Rabbit
Immunogen Synthetic peptide within Human ADAM10 aa 715-748.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102, 11487, 29650
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.