Learn More
ACROBiosystems Recombinant Protein;Mouse ADAM17 / TACE / CD156b Protein;HEK293;50UG;AD7,ADAM17,TACE,CD156b
Supplier:  ACROBiosystems AD7M52H150UG
Encompass_Preferred
Recombinant Protein;Mouse ADAM17 / TACE / CD156b Protein;50UG;Mouse ADAM17, His Tag (AD7-M52H1) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - Asp 563 (Accession # Q9Z0F8).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 63.4 kDa. The protein migrates as 19 kDa and 20 kDa(Propeptide),55-66 kDa (Mature-ADAM17) and 75-94 kDa(Pro-ADAM17) under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8769
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.